Learn More
Biomatik Corporation Rat Immunoglobulin E, IgE ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39214
Encompass_Preferred
Rat Immunoglobulin E, Ige Elisa Kit for detecting Immunoglobulin E, Ige in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8897