Learn More
Biomatik Corporation Rat inducible nitric oxide synthase, iNOS ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39220
Encompass_Preferred
Rat Inducible Nitric Oxide Synthase, Inos Elisa Kit for detecting Nitric Oxide Synthase 2, Inducible in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50IU/ml. Sensitivity: 0.195IU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q06518; Nos2. Target Synonyms: Nos2; Nitric oxide synthase; inducible; EC 1.14.13.39; Inducible NO synthase; Inducible NOS; iNOS; NOS type II; Peptidyl-cysteine S-nitrosylase NOS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8899