Learn More
Biomatik Corporation Rat inositol 1, 4, 5, -trisphosphate (IP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39227
Encompass_Preferred
Rat Inositol 1, 4, 5, -Trisphosphate (Ip3) Elisa Kit for detecting Inositol 1, 4, 5, -Trisphosphate(Ip3) in serum, plasma, tissue, homogenates, cell culture supernates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 5-1000pg/ml. Sensitivity: 2.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8901