Learn More
Biomatik Corporation Rat Interleukin 18, IL-18 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39260
Encompass_Preferred
Rat Interleukin 18, Il-18 Elisa Kit for detecting Interleukin 18 (Interferon-Gamma-Inducing Factor) in serum, plasma, cell culture supernates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P97636; Il18. Target Synonyms: Il18; Igif; Interleukin-18; IL-18; Interferon gamma-inducing factor; IFN-gamma-inducing factor; Interleukin-1 gamma; IL-1 gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8908