Learn More
Biomatik Corporation Rat Interleukin 1, IL-1 ELISA Kit, 96 tests
Catalog No. 501498909
Encompass_Preferred
Rat Interleukin 1Α , Il-1Α Elisa Kit for detecting Interleukin 1, Alpha in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04822; IL1A. Target Synonyms: IL1A; Interleukin-1 alpha; IL-1 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8909 Supplier Biomatik Corporation Supplier No. EKC39261
Rat Interleukin 1Α , Il-1Α Elisa Kit for detecting Interleukin 1, Alpha in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04822; IL1A. Target Synonyms: IL1A; Interleukin-1 alpha; IL-1 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.