Learn More
Biomatik Corporation Rat Interleukin 1, IL-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39262
Encompass_Preferred
Rat Interleukin 1Β, Il-1Β Elisa Kit for detecting Interleukin 1, Beta in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q63264; Il1b. Target Synonyms: Il1bInterleukin-1 beta; IL-1 beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8910