Learn More
Biomatik Corporation Rat Leukotriene B4, LT-B4 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39340
Encompass_Preferred
Rat Leukotriene B4, Lt-B4 Elisa Kit for detecting Leukotriene B4, Lt-B4 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8918