Learn More
Biomatik Corporation Rat lipolysaccharide binding protein, LBP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39345
Encompass_Preferred
Rat Lipolysaccharide Binding Protein, Lbp Elisa Kit for detecting Lipopolysaccharide Binding Protein in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q63313; Lbp. Target Synonyms: LbpLipopolysaccharide-binding protein; LBP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8919