Learn More
Biomatik Corporation Rat malonyl coenzyme A ELISA Kit, 96 tests
Catalog No. 501498924
Encompass_Preferred
Rat Malonyl Coenzyme A Elisa Kit for detecting Malonyl Coenzyme A in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.25-20ng/ml. Sensitivity: 0.625ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8924 Supplier Biomatik Corporation Supplier No. EKC39374
Rat Malonyl Coenzyme A Elisa Kit for detecting Malonyl Coenzyme A in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.25-20ng/ml. Sensitivity: 0.625ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.