Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 14, MMP-14 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39387
Encompass_Preferred
Rat Matrix Metalloproteinase 14, Mmp-14 Elisa Kit for detecting Matrix Metallopeptidase 14 (Membrane-Inserted) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 46.88-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q10739 ; Mmp14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8926