Learn More
Biomatik Corporation Rat matrix metalloproteinase 7, MMP-7 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39391
Encompass_Preferred
Rat Matrix Metalloproteinase 7, Mmp-7 Elisa Kit for detecting Matrix Metallopeptidase 7 (Matrilysin, Uterine) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P50280; Mmp7. Target Synonyms: Mmp7; Mmp-7Matrilysin; EC 3.4.24.23; Matrin; Matrix metalloproteinase-7; MMP-7; Pump-1 protease; Uterine metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8928