Learn More
Biomatik Corporation Rat Neurofilament light polypeptide (NEFL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39470
Encompass_Preferred
Rat Neurofilament Light Polypeptide (Nefl) Elisa Kit for detecting Neurofilament, Light Polypeptide in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19527; Nefl. Target Synonyms: Nefl; Nf68; Nfl; Neurofilament light polypeptide; NF-L; 68 kDa neurofilament protein; Neurofilament triplet L protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8945