Learn More
Biomatik Corporation Rat Paraoxonase 1 (PON1) ELISA Kit, 96 tests
Catalog No. 501498956
Encompass_Preferred
Rat Paraoxonase 1 (Pon1) Elisa Kit for detecting Paraoxonase 1 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000uU/ml. Sensitivity: 3.9uU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55159; Pon1. Target Synonyms: Pon1; Pon; Serum paraoxonase/arylesterase 1; PON 1; EC 3.1.1.2; EC 3.1.1.81; EC 3.1.8.1; Aromatic esterase 1; A-esterase 1; Serum aryldialkylphosphatase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8956 Supplier Biomatik Corporation Supplier No. EKC39527
Rat Paraoxonase 1 (Pon1) Elisa Kit for detecting Paraoxonase 1 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000uU/ml. Sensitivity: 3.9uU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55159; Pon1. Target Synonyms: Pon1; Pon; Serum paraoxonase/arylesterase 1; PON 1; EC 3.1.1.2; EC 3.1.1.81; EC 3.1.8.1; Aromatic esterase 1; A-esterase 1; Serum aryldialkylphosphatase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.