Learn More
Biomatik Corporation Rat Prostaglandin-H2 D-isomerase (PTGDS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39595
Encompass_Preferred
Rat Prostaglandin-H2 D-Isomerase (Ptgds) Elisa Kit for detecting Prostaglandin D2 Synthase 21Kda (Brain) in serum, plasma, cerebrospinal fluid (csf), tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 9.37-600ng/ml. Sensitivity: 2.34ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22057; Ptgds. Target Synonyms: Ptgds; Prostaglandin-H2 D-isomerase; EC 5.3.99.2; Glutathione-independent PGD synthase; Lipocalin-type prostaglandin-D synthase; Prostaglandin-D2 synthase; PGD2 synthase; PGDS; PGDS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8968