Learn More
Biomatik Corporation Rat Protein CYR61 (CYR61) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39600
Encompass_Preferred
Rat Protein Cyr61 (Cyr61) Elisa Kit for detecting Cysteine-Rich, Angiogenic Inducer, 61 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9ES72; Cyr61. Target Synonyms: Ccn1; Cyr61; Igfbp10CCN family member 1; Cellular communication network factor 1; Cysteine-rich angiogenic inducer 61; Insulin-like growth factor-binding protein 10; IBP-10; IGF-binding protein 10; IGFBP-10; Protein CYR61. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8970