Learn More
Biomatik Corporation Rat relaxin 1 (RLN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39640
Encompass_Preferred
Rat Relaxin 1 (Rln1) Elisa Kit for detecting Relaxin 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.4-150pg/ml. Sensitivity: 0.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01347; Rln1. Target Synonyms: Rln1; Rln; Prorelaxin 1 [Cleaved into: Relaxin B chain; Relaxin A chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8976