Learn More
Biomatik Corporation Rat Secretin (Sct) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39672
Encompass_Preferred
Rat Secretin (Sct) Elisa Kit for detecting Secretin (Sct) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11384; Sct. Target Synonyms: Sct; Secretin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8981