Learn More
Biomatik Corporation Rat Serotonin Receptor 2A/5-hydroxytryptamine Receptor 2A (5-HT-2A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39684
Encompass_Preferred
Rat Serotonin Receptor 2A/5-Hydroxytryptamine Receptor 2A (5-Ht-2A) Elisa Kit for detecting 5-Hydroxytryptamine (Serotonin) Receptor 2A in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P14842; Htr2a. Target Synonyms: Htr2a; Htr2; 5-hydroxytryptamine receptor 2A; 5-HT-2; 5-HT-2A; Serotonin receptor 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8982