Learn More
Biomatik Corporation Rat soluble fibrin monomer complex (SFMC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39705
Encompass_Preferred
Rat Soluble Fibrin Monomer Complex (Sfmc) Elisa Kit for detecting Soluble Fibrin Monomer Complex(Sfmc) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8987