Learn More
Biomatik Corporation Rat Thromboxane B2, TXB2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39767
Encompass_Preferred
Rat Thromboxane B2, Txb2 Elisa Kit for detecting Thromboxane B2, Txb2 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.9-250pg/ml. Sensitivity: 0.97pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8996