Learn More
Biomatik Corporation Rat tumor necrosis factor-related apoptosis inducing ligand, TRAIL ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39840
Encompass_Preferred
Rat Tumor Necrosis Factor-Related Apoptosis Inducing Ligand, Trail Elisa Kit for detecting Tumor Necrosis Factor-Related Apoptosis Inducing Ligand, Trail in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P50591; TNFSF10. Target Synonyms: Apo 2 ligand; APO 2L; Apo-2 ligand; Apo-2L; APO2L; CD253; CD253 antigen; Chemokine tumor necrosis factor ligand superfamily member 10; Protein TRAIL; TL2; TNF Related Apoptosis Inducing Ligand; TNF related apoptosis inducing ligand TRAIL; TNF-related apoptosis-inducing ligand; TNF10_HUMAN; Tumor necrosis factor (ligand) family member 10; Tumor Necrosis Factor (ligand) Superfamily Member 10; Tumor necrosis factor apoptosis inducing ligand splice variant delta; Tumor necrosis factor ligand superfamily member 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9004