Learn More
Biomatik Corporation Rat Undercarboxylated Osteocalcin (ucOC) ELISA Kit, 96 tests
Catalog No. 501499005
Encompass_Preferred
Rat Undercarboxylated Osteocalcin (Ucoc) Elisa Kit for detecting Undercarboxylated Osteocalcin (Ucoc) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9005 Supplier Biomatik Corporation Supplier No. EKC39852
Rat Undercarboxylated Osteocalcin (Ucoc) Elisa Kit for detecting Undercarboxylated Osteocalcin (Ucoc) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.