Learn More
Biomatik Corporation Rat Vascular endothelial cell growth factor receptor 1, VEGFR-1/Flt1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39865
Encompass_Preferred
Rat Vascular Endothelial Cell Growth Factor Receptor 1, Vegfr-1/Flt1 Elisa Kit for detecting Fms-Related Tyrosine Kinase 1 (Vascular Endothelial Growth Factor/Vascular Permeability Factor Receptor) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P53767; Flt1. Target Synonyms: Flt1; Flt-1; Vegfr1; Vascular endothelial growth factor receptor 1; VEGFR-1; EC 2.7.10.1; Fms-like tyrosine kinase 1; FLT-1; Tyrosine-protein kinase receptor FLT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9008