Learn More
Biomatik Corporation Rat Vitamin D3 receptor (VDR) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKC39877
Rat Vitamin D3 Receptor (Vdr) Elisa Kit for detecting Vitamin D (1, 25- Dihydroxyvitamin D3) Receptor in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13053; Vdr. Target Synonyms: Vdr; Nr1i1Vitamin D3 receptor; VDR; 1; 25-dihydroxyvitamin D3 receptor; Nuclear receptor subfamily 1 group I member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.