Learn More
Biomatik Corporation Rat von Willebrand Factor, vWF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39882
Encompass_Preferred
Rat Von Willebrand Factor, Vwf Elisa Kit for detecting Von Willebrand Factor in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q62935; Vwf. Target Synonyms: Vwf; von Willebrand factor; vWF; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9011