Learn More
Biomatik Corporation Rat Fibronectin, FN ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40044
Encompass_Preferred
Rat Fibronectin, Fn Elisa Kit for detecting Fibronectin 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04937; Fn1. Target Synonyms: Fn1Fibronectin; FN) [Cleaved into: Anastellin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9031