Learn More
Biomatik Corporation Mouse Interferon , IFN- ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKC40057
Mouse Interferon Γ , Ifn-Γ Elisa Kit for detecting Interferon, Gamma in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01580; Ifng. Target Synonyms: Ifng; Interferon gamma; IFN-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.