Learn More
Biomatik Corporation Human Interleukin 8, IL-8 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40086
Encompass_Preferred
The Human Interleukin 8, IL-8 ELISA Kit allows for quantitative determination interleukin 8 in serum, cell culture supernates, saliva, urine, cerebrospinal fluid (CSF), tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.110 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P10145. Restriction: For Research Use Only.
Catalog No. 50-149-9055