Learn More
Biomatik Corporation Human monocyte chemotactic protein 1/monocyte chemotactic and activating factor, MCP-1/MCAF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40095
Encompass_Preferred
The Human monocyte chemotactic protein 1/monocyte chemotactic and activating factor, MCP-1/MCAF ELISA Kit allows for quantitative determination of chemokine (C-C motif) ligand 2 in serum, plasma, urine, saliva, cerebrospinal fluid (CSF), ascitic fluid, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 30.184 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P13500. Restriction: For Research Use Only.
Catalog No. 50-149-9058