Learn More
Biomatik Corporation Human matrix metalloproteinase 3/stromelysin 1 (MMP3/STR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40103
Encompass_Preferred
The Human matrix metalloproteinase 3/stromelysin 1 (MMP3/STR1) ELISA Kit allows for quantitative determination of matrix metallopeptidase 3 (stromelysin 1, progelatinase) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P08254. Restriction: For Research Use Only.
Catalog No. 50-149-9061