Learn More
Biomatik Corporation Mouse Stromal cell derived factor 1, SDF-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40116
Encompass_Preferred
Mouse Stromal Cell Derived Factor 1Β, Sdf-1Β Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 12 (Stromal Cell-Derived Factor 1) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40224; Cxcl12. Target Synonyms: Cxcl12; Sdf1; Stromal cell-derived factor 1; SDF-1; 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; TPAR1; C-X-C motif chemokine 12; Pre-B cell growth-stimulating factor; PBSF; Thymic lymphoma cell-stimulating factor; TLSF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9067