Learn More
Biomatik Corporation Human Tumor necrosis factor , TNF- ELISA Kit, 96 tests
Catalog No. 501499070
Encompass_Preferred
Human Tumor Necrosis Factor Α, Tnf-Α Elisa Kit for detecting Tumor Necrosis Factor (Tnf Superfamily, Member 2) in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01375; TNF. Target Synonyms: APC1; APC1 protein; Cachectin; DIF; Differentiation inducing factor; Macrophage cytotoxic factor; Tnf; TNF superfamily member 2; TNF superfamily; member 2; TNF; macrophage derived; TNF; monocyte derived; TNF-a; TNF-alpha; TNFA; TNFA_HUMAN; TNFSF2; Tumor necrosis factor (TNF superfamily member 2); Tumor necrosis factor alpha; Tumor necrosis factor; Tumor necrosis factor ligand superfamily member 2; Tumor Necrosis Factor; Membrane Form; Tumor necrosis factor; soluble form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9070 Supplier Biomatik Corporation Supplier No. EKC40121
Human Tumor Necrosis Factor Α, Tnf-Α Elisa Kit for detecting Tumor Necrosis Factor (Tnf Superfamily, Member 2) in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01375; TNF. Target Synonyms: APC1; APC1 protein; Cachectin; DIF; Differentiation inducing factor; Macrophage cytotoxic factor; Tnf; TNF superfamily member 2; TNF superfamily; member 2; TNF; macrophage derived; TNF; monocyte derived; TNF-a; TNF-alpha; TNFA; TNFA_HUMAN; TNFSF2; Tumor necrosis factor (TNF superfamily member 2); Tumor necrosis factor alpha; Tumor necrosis factor; Tumor necrosis factor ligand superfamily member 2; Tumor Necrosis Factor; Membrane Form; Tumor necrosis factor; soluble form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.