Learn More
Biomatik Corporation Human prostate specific antigen, PSA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40133
Encompass_Preferred
The Human prostate specific antigen, PSA ELISA Kit allows for quantitative determination of kallikrein-related peptidase 3 in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07288. Restriction: For Research Use Only.
Catalog No. 50-149-9076