Learn More
Biomatik Corporation Dog Transforming growth factor β1, TGF-β1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40142
Encompass_Preferred
The Dog Transforming growth factor β1, TGF-β1 ELISA Kit allows for quantitative determination of transforming growth factor, beta 1 in serum, plasma. Species Reactivity: Dog. Detection Range: 0.781 ng/mL-50 ng/mL. Sensitivity: 0.747 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P54831. Restriction: For Research Use Only.
Catalog No. 50-149-9077