Learn More
Biomatik Corporation Mouse neutrophil elastase, NE ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40144
Encompass_Preferred
Mouse Neutrophil Elastase, Ne Elisa Kit for detecting Elastase 2, Neutrophil in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q3UP87; Elane. Target Synonyms: Elane; Ela2Neutrophil elastase; EC 3.4.21.37; Elastase-2; Leukocyte elastase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9078