Learn More
Biomatik Corporation Mouse Interferon , IFN-/IFNB ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40162
Encompass_Preferred
Mouse Interferon Β, Ifn-Β/Ifnb Elisa Kit for detecting Interferon, Beta 1, Fibroblast in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01575; Ifnb1. Target Synonyms: Ifnb1; Ifb; IfnbInterferon beta; IFN-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9080