Learn More
Biomatik Corporation Rat Testosterone, T ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40191
Encompass_Preferred
Rat Testosterone, T Elisa Kit for detecting Testosterone, T in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.13-25.6ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9085