Learn More
Biomatik Corporation Rat Estradiol, E2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40197
Encompass_Preferred
Rat Estradiol, E2 Elisa Kit for detecting Estradiol, E2 in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 40-1000pg/ml. Sensitivity: 40pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9090