Learn More
Biomatik Corporation Pig Interleukin 1, IL-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40216
Encompass_Preferred
Pig Interleukin 1Β, Il-1Β Elisa Kit for detecting Interleukin 1, Beta in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 3.9-250pg/ml. Sensitivity: 0.975pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P26889; IL1B. Target Synonyms: IL1BInterleukin-1 beta; IL-1 beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9099