Learn More
Biomatik Corporation Pig tissue inhibitors of metalloproteinase 1, TIMP-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40222
Encompass_Preferred
Pig Tissue Inhibitors Of Metalloproteinase 1, Timp-1 Elisa Kit for detecting Timp Metallopeptidase Inhibitor 1 in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.8-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35624; TIMP1. Target Synonyms: TIMP1; Metalloproteinase inhibitor 1; Tissue inhibitor of metalloproteinases 1; TIMP-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9104