Learn More
Biomatik Corporation Pig Immunoglobulin G (IgG) ELISA Kit, 96 tests
Catalog No. 501499105
Encompass_Preferred
Pig Immunoglobulin G (Igg) Elisa Kit for detecting Immunoglobulin G(Igg) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.586-150ug/ml. Sensitivity: 0.467ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9105 Supplier Biomatik Corporation Supplier No. EKC40224
Pig Immunoglobulin G (Igg) Elisa Kit for detecting Immunoglobulin G(Igg) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.586-150ug/ml. Sensitivity: 0.467ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.