Learn More
Biomatik Corporation Pig endothelin 1, ET-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40226
Encompass_Preferred
Pig Endothelin 1, Et-1 Elisa Kit for detecting Endothelin 1 in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09558; EDN1. Target Synonyms: EDN1; Endothelin-1; ET-1; Preproendothelin-1; PPET1) [Cleaved into: Big endothelin-1]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9106