Learn More
Biomatik Corporation Pig brain natriuretic peptide, BNP ELISA Kit, 96 tests
Catalog No. 501499107
Encompass_Preferred
Pig Brain Natriuretic Peptide, Bnp Elisa Kit for detecting Brain Natriuretic Peptide, Bnp in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07634; NPPB. Target Synonyms: NPPBNatriuretic peptides B; Gamma-brain natriuretic peptide) [Cleaved into: Brain natriuretic peptide 32; BNP-32); Brain natriuretic peptide 26; BNP-26)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9107 Supplier Biomatik Corporation Supplier No. EKC40227
Pig Brain Natriuretic Peptide, Bnp Elisa Kit for detecting Brain Natriuretic Peptide, Bnp in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07634; NPPB. Target Synonyms: NPPBNatriuretic peptides B; Gamma-brain natriuretic peptide) [Cleaved into: Brain natriuretic peptide 32; BNP-32); Brain natriuretic peptide 26; BNP-26)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.