Learn More
Biomatik Corporation Rabbit C-Reactive Protein, CRP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40233
Encompass_Preferred
Rabbit C-Reactive Protein, Crp Elisa Kit for detecting C-Reactive Protein, Pentraxin-Related in serum, plasma. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 7.8-2000pg/ml. Sensitivity: 2pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02742; CRP. Target Synonyms: CRP; PTX1C-reactive protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9109