Learn More
Biomatik Corporation Rat adrenocorticotropic hormone (ACTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40237
Encompass_Preferred
Rat Adrenocorticotropic Hormone (Acth) Elisa Kit for detecting Adrenocorticotropic Hormone (Acth) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.25-50pg/ml. Sensitivity: 1.25pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9112