Learn More
Biomatik Corporation rat prolactin, PRL ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40238
Encompass_Preferred
Rat Prolactin, Prl Elisa Kit for detecting Prolactin in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.125-50ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P01237; Prl. Target Synonyms: Prl; Prolactin; PRL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9113