Learn More
Biomatik Corporation Human Resistin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40240
Encompass_Preferred
Human Resistin Elisa Kit for detecting Resistin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9HD89; RETN. Target Synonyms: Adipose tissue specific secretory factor; Adipose tissue-specific secretory factor; ADSF; C/EBP epsilon regulated myeloid specific secreted cysteine rich protein; C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 1; C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; CG5403; Cysteine rich secreted protein A12 alpha like 2; Cysteine rich secreted protein FIZZ3; Cysteine-rich secreted protein A12-alpha-like 2; Cysteine-rich secreted protein FIZZ3; dri; FIZZ 3; FIZZ3; Found in inflammatory zone 3; HXCP 1; HXCP1; MGC126603; MGC126609; PRO1199; Resistin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9115