Learn More
Biomatik Corporation rat Resistin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40241
Encompass_Preferred
Rat Resistin Elisa Kit for detecting Resistin in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8K4J7; Retn. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9116