Learn More
Biomatik Corporation Human Progesterone, PROG ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40274
Encompass_Preferred
The Human Progesterone, PROG ELISA Kit allows for quantitative determination of Progesterone, PROG in serum, plasma. Species Reactivity: Human. Detection Range: 1 ng/mL-30 ng/mL. Sensitivity: 0.2 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9134