Learn More
Biomatik Corporation Rat Interleukin 9, IL-9 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40281
Encompass_Preferred
Rat Interleukin 9, Il-9 Elisa Kit for detecting Interleukin 9, Il-9 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: D4A8I9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9140